HiLoad Superdex columns, 75 PG
About this item
Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component
- High resolution with short run times and good recovery
- High capacity at high flow rates
- Separations can be scaled up to production levels
- Prepacked for convenience and reproducibility
- 1/16" fittings for convenient connections to ÄKTA design systems
Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr < 10 000 (Superdex 30 pg), Mr 3 000 to 70 000 (Superdex 75 pg), and Mr 10 000 600 000 (Superdex 200 pg). The medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions.
Delivery information: Includes dismantling tool, support screen, net ring (10 μm), O-ring, stop plugs and instructions.
2 Options Available Below
Product Details & Documents
Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component
- High resolution with short run times and good recovery
- High capacity at high flow rates
- Separations can be scaled up to production levels
- Prepacked for convenience and reproducibility
- 1/16" fittings for convenient connections to ÄKTA design systems
Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr < 10 000 (Superdex 30 pg), Mr 3 000 to 70 000 (Superdex 75 pg), and Mr 10 000 600 000 (Superdex 200 pg). The medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions.
Delivery information: Includes dismantling tool, support screen, net ring (10 μm), O-ring, stop plugs and instructions.
Documents
Certificates
Search by Lot Number28989333_DATA FILE 28989334_DATA FILE
Datasheet
28989333_HAND BOOK 28989333_HAND BOOK 1 28989333_INSTRUCTIONS 28989333_INSTRUCTIONS 1 28989334_HAND BOOK
Manual
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications for Product Family
Specifications
- Catalog No:
- 28-9893-33
- 28-9893-34
- Max. pressure:
- 3 bar
- 3 bar
- pH range:
- 3 - 12
- 3 - 12
- Storage:
- 4 to 30 °C, 20% Ethanol
- 4 to 30 °C, 20% Ethanol
Specifications
Product Family Options
Product Information
- Column hardwareParticle SizeBed dimensionsCapacitySample volumeFlow rateFlow velocityAvailabilityPrice
- 28-9893-33HiLoad™ 16/600 Superdex™ 75 PGHiLoad™ 16/600 Superdex™ 75 PGGlass34 µm16×600 mm120 ml≤ 5 ml<1,0 ml/min10 - 50 cm/hLog in for availability1 * 1 ST2 930,00 €Specifications:
More
Specifications
Cat. No.28-9893-33Max. pressure3 barpH range3 - 12Storage4 to 30 °C, 20% Ethanol - 28-9893-34HiLoad™ 26/600 Superdex™ 75 PGHiLoad™ 26/600 Superdex™ 75 PGGlass34 µm26×600 mm320 ml≤ 13 ml<2,6 ml/min10 - 50 cm/hLog in for availability1 * 1 ST4 400,00 €Specifications:
More
Specifications
Cat. No.28-9893-34Max. pressure3 barpH range3 - 12Storage4 to 30 °C, 20% Ethanol
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



