ContactUsLinkComponent
Syringe filters, prefilter, sterile, GD/X™, Whatman™
Syringe filters, prefilter, sterile, GD/X™, Whatman™
Certificates
undefined
Syringe filters, prefilter, sterile, GD/X™, Whatman™
Certificates
Specifications
  • Catalog No:
  • 514-0538
  • 514-0541
  • 514-0542
  • 514-0539
  • 514-0540
  • 514-0537
  • 516-6280
  • Dimensions:
  • 21,6×29,8 mm
  • 21,6×29,8 mm
  • 21,6×29,8 mm
  • 21,6×29,8 mm
  • 21,6×29,8 mm
  • 21,6×29,8 mm
  • 21,6×29,8 mm
  • Max. operating pressure:
  • 5,2 bar
  • 5,2 bar
  • 5,2 bar
  • 5,2 bar
  • 5,2 bar
  • 5,2 bar
  • 5,2 bar
  • Weight:
  • 3 g
  • 3 g
  • 3 g
  • 3 g
  • 3 g
  • 3 g
  • 3 g