Order Entry
Austria
ContactUsLinkComponent
Universal reverse primer
Universal reverse primer
Catalog # AGLS600037
Supplier:  AGILENT
CAS Number:  
Universal reverse primer
Catalog # AGLS600037
Supplier:  AGILENT
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Universal reverse primer
  • Seq:
    02
  • No. of reactions:
    50

Specifications

About this item

The universal reverse primer serves as the downstream primer in the qPCR reaction by annealing to a universal sequence tag that is added to the cDNA during the reverse transcription protocol.

Applications: PCR
Number of Reactions: 3.125 μM
Physical Form: Liquid
Reactions per Kit: 300
Volume: 200 µL