Order Entry
Austria
ContactUsLinkComponent
Brilliant II SYBR® master mix with ROX
Brilliant II SYBR® master mix with ROX
Catalog # AGLS600830
Supplier:  AGILENT
CAS Number:  
Brilliant II SYBR® master mix with ROX
Catalog # AGLS600830
Supplier:  AGILENT
CAS Number:  

Specifications

  • Pk:
    1
  • Application:
    qPCR dye
  • Description:
    Brilliant II SYBR Green Low ROX QPCR Master Mix
  • Seq:
    4
  • Details:
    High performance, ultra sensitive, brighter and faster next generation SYBR(R) Green QPCR master mix reagent with ROX reference dye for reliable quantification across a wide range of targets and templates
  • No. of reactions:
    400 reactions

Specifications

About this item

Brilliant II SYBR® QPCR and QRT-PCR master mix kits with high/low Rox offer superior sensitivity for improved quantification and reproducibility.

  • Improved performance with earlier Ct detection across a wide dynamic range
  • Increased sensitivity of detection to low copy numbers

It also shows more consistent performance across a wide range of template concentrations and different targets.

Caution: For research use only. Not for use in diagnostic procedures.