SymLine® FLEX Starter Sets
About this item
The flexible waste disposal system for HPLC workstations. Requiring only a few basic elements, this modular system can be adapted to suit virtually any workplace.
- Simple installation, with table or wall mounting possible
- Do-It-Yourself modular system is designed for its suitability for any workplace
- A minimum of components, with many applications
- Can be flexibly integrated at any time; react quickly to any changes in the laboratory
- Whether adding additional workplaces or a reinstallation at another location, the system can be made to fit
- Anti-static and resistant to solvents
- Electrically conductive plastic ensures secure grounding/earthing, to avoid danger of ignition
The SymLine® FLEX modular system fits snugly into virtually any HPLC workplace. It now only requires a single waste disposal container. No other awkwardly placed waste containers need be used within the working area. The flexible table installation allows for connections to existing equipment to be made at any desired place. The resultant height then gives the required gradient, leading to a safe downward flow of liquid. Table mounting is possible for various tabletop thicknesses, and the required attachments can be installed at the tabletop without the use of any tools. The pipe system may be lengthened or shortened, according to specific needs. All the pipework is intercompatible and easily interconnected. Central collection point for solvent waste eliminates working area obstructions; liquids are safely collected at one central point.
The resultant height then gives the required gradient, leading to a safe downward flow of liquid. A table mounting is possible for various tabletop thicknesses, and the required attachments can be installed at the tabletop without the use of any tools. The pipe system may be lengthened or shortened, according to specific wishes. All the pipework is intercompatible and easily interconnected.
Ordering information: Premium Starter Set includes:
1× SCTV106688 - Basic pipe
1× SCTV106423 - Blind plug for pipe
1× SCTV106568 - SymLine Flexube
1× SCTV106475 - Shut-off, angled
1× SCTV490336 - Exhaust filter M, V3.0, economy package
1× SCTV107624 - Offset adapter 90° for exhaust filter
1× SCTV106480 - Safety waste cap, S60/61
2x SCTV107953 - Canister 10 L, S60/61
1× SCTV117986 - Collecting tray with base insert
1× SCTV108176 - Grounding cable
1 x SCTV108050 - Signal cable for electronic level control
1 x SCTV108087 - Signal box T1 - EU
Standard Starter Set includes:
1× SCTV106688 - Basic 32 pipe
1× SCTV106423 - Blind plug for 32 pipe
1× SCTV106568 - SymLine FlexTube
1× SCTV106475 - Shut-off, angled
1× SCTV490336 - Exhaust filter M, V3.0, economy package
1× SCTV306482 - Safety waste cap, S60/61
2x SCTV108042 - Canister 10 L, S60/61
1× SCTV117986 - Collecting tray with base insert
1× SCTV108176 - Grounding cable
4 Options Available Below
Product Details & Documents
The flexible waste disposal system for HPLC workstations. Requiring only a few basic elements, this modular system can be adapted to suit virtually any workplace.
- Simple installation, with table or wall mounting possible
- Do-It-Yourself modular system is designed for its suitability for any workplace
- A minimum of components, with many applications
- Can be flexibly integrated at any time; react quickly to any changes in the laboratory
- Whether adding additional workplaces or a reinstallation at another location, the system can be made to fit
- Anti-static and resistant to solvents
- Electrically conductive plastic ensures secure grounding/earthing, to avoid danger of ignition
The SymLine® FLEX modular system fits snugly into virtually any HPLC workplace. It now only requires a single waste disposal container. No other awkwardly placed waste containers need be used within the working area. The flexible table installation allows for connections to existing equipment to be made at any desired place. The resultant height then gives the required gradient, leading to a safe downward flow of liquid. Table mounting is possible for various tabletop thicknesses, and the required attachments can be installed at the tabletop without the use of any tools. The pipe system may be lengthened or shortened, according to specific needs. All the pipework is intercompatible and easily interconnected. Central collection point for solvent waste eliminates working area obstructions; liquids are safely collected at one central point.
The resultant height then gives the required gradient, leading to a safe downward flow of liquid. A table mounting is possible for various tabletop thicknesses, and the required attachments can be installed at the tabletop without the use of any tools. The pipe system may be lengthened or shortened, according to specific wishes. All the pipework is intercompatible and easily interconnected.
Ordering information: Premium Starter Set includes:
1× SCTV106688 - Basic pipe
1× SCTV106423 - Blind plug for pipe
1× SCTV106568 - SymLine Flexube
1× SCTV106475 - Shut-off, angled
1× SCTV490336 - Exhaust filter M, V3.0, economy package
1× SCTV107624 - Offset adapter 90° for exhaust filter
1× SCTV106480 - Safety waste cap, S60/61
2x SCTV107953 - Canister 10 L, S60/61
1× SCTV117986 - Collecting tray with base insert
1× SCTV108176 - Grounding cable
1 x SCTV108050 - Signal cable for electronic level control
1 x SCTV108087 - Signal box T1 - EU
Standard Starter Set includes:
1× SCTV106688 - Basic 32 pipe
1× SCTV106423 - Blind plug for 32 pipe
1× SCTV106568 - SymLine FlexTube
1× SCTV106475 - Shut-off, angled
1× SCTV490336 - Exhaust filter M, V3.0, economy package
1× SCTV306482 - Safety waste cap, S60/61
2x SCTV108042 - Canister 10 L, S60/61
1× SCTV117986 - Collecting tray with base insert
1× SCTV108176 - Grounding cable
Product Family Options
Sets
Product Information
- AvailabilityPrice
- SCTV106694Starter set, premium
Mounting
Product Information
- AvailabilityPrice
- SCTV106691Table mount, PEDH-EC
Piping
Product Information
- AvailabilityPrice
- SCTV106690Pipe extension 1, 600 mm, PEDH-EC
- SCTV106732Pipe extension 4, 1200 mm, 1× GL 45 (m), PEDH-EC, 32 mm int.ØPipe extension 4, 1200 mm, 1× GL 45 (m), PEDH-EC, 32 mm int.ØLog in for availability1 * 1 ST225,23 €
Piping
Product Information
- Catalog #ThreadDescriptionAvailabilityPrice
Filter
Product Information
- Catalog #DescriptionAvailabilityPrice
- Exhaust filter M, V3.0 with splash guard and change indicator, 6 month service lifeLog in for availability1 * 2 ST171,54 €
Safety waste caps
Product Information
- Catalog #ThreadMaterialConductivityDescriptionAvailabilityPrice
Canister
Product Information
- Catalog #ThreadCapacityW×D×HTypeDescriptionColourAvailabilityPrice
Tray
Product Information
- Catalog #DescriptionTypeW×D×H ext.W×D×H int.ColourAvailabilityPrice
- Collecting tray with base insert, PEDH-EC materialElectrically conductive, with base insert365×490×240 mm295×415×200 mmBlackLog in for availability1 * 1 ST303,49 €
Signal box
Product Information
- Catalog #DescriptionAvailabilityPrice