Order Entry
Austria
ContactUsLinkComponent
 
Affinity chromatography columns, HiTrap™ protein G HP
 :  Cytiva
 
undefined
Affinity chromatography columns, HiTrap™ protein G HP
 :  Cytiva
 

 

  • Catalog No:
  • 17-0405-03
  • 17-0404-03
  • 17-0405-01
  • 17-0404-01
  • 29-0485-81
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 9
  • 3 - 9
  • 3 - 9
  • 3 - 9
  • 3 - 9
  • Storage:
  • 2 to 8 °C, 20% Ethanol
  • 2 to 8 °C, 20% Ethanol
  • 2 to 8 °C, 20% Ethanol
  • 2 to 8 °C, 20% Ethanol
  • 2 to 8 °C, 20% Ethanol

 

Frequently Bought Together