Orders LinkContactUsLinkComponent
Affinity protein expression and purification systems
Affinity protein expression and purification systems
Supplier:   STRATAGENE
Affinity protein expression and purification systems
Supplier:  STRATAGENE
Certificates
Affinity protein expression and purification systems
2 Options Available Below
Product Details & Documents
The affinity protein expression and purification system provides a simple, gentle and effective method for high-level protein expression and purification.
  • pCAL vectors derived from T7 RNA polymerase-based pET vectors
  • Novel purification tag combines small size, gentle elution and high-affinity binding
  • Easy tag removal resulting in native protein
Delivery information: The complete system includes pCAL expression vectors containing a calmodulin-binding peptide tag and calmodulin affinity resin (available separately).
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Product Information
  • Details
    Pk
    Availability
    Price
  • AGLS214303
    Calmodulin affinity resin
    Calmodulin affinity resin
    Calmodulin binding peptide (CBP) affinity resin
    10 mL
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - 470,00 €
    470,00 €
    1 * 10 mL
       470,00 €
  • STRA214303
    Calmodulin affinity resin
    Calmodulin affinity resin
    Calmodulin binding peptide (CBP) affinity resin
    10 mL
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL
    Specifications:
    Marketsource
    Marketsource
This product has been blocked by your organization. Please contact your purchasing department for more information