ContactUsLinkComponent
VWR®, Cell culture media, Minimum Essential Medium (MEM)
VWR®, Cell culture media, Minimum Essential Medium (MEM)
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Cell culture media, Minimum Essential Medium (MEM)
Supplier:  VWR Chemicals
Certificates
Specifications
  • Calcium:
    Yes
  • Magnesium:
    Yes
  • HEPES:
    No
  • Sodium bicarbonate:
    Yes
  • Sodium pyruvate:
    No
  • Phenol red:
    Yes
  • Stable glutamine:
    No
  • L-Glutamine:
    No
  • Media Base:
    MEM/EBSS - Minimum Essential Medium with Earle's Balanced Salts
  • Cat. No.:
    392-0424