GC columns, J&W HP-50+
: Agilent
HP-50+ is excellent for confirmational analyses of polar compounds such as chlorinated pesticides, herbicides, and basic drugs.
- (50%-Phenyl)-methylpolysiloxane
- Mid-polarity - slightly less polar than DB-17
- Excellent for confirmational analyses
- Bonded and cross-linked
- Solvent rinsable
J&W HP 50+ is a (50% phenyl) methylpolysiloxane phase which is excellent for confirmational analysis. These mid-polarity phase columns are generally used for analysis of polar compounds such as chlorinated pesticides, herbicides, and basic drugs. They are often used in conjunction with a second column to provide confirmational information. Additionally, the phase ratios of the 0,32 mm series are maintained in the 0,20 mm series, simplifying transfer of methods to GC/MS systems.
Frequently Bought Together