ContactUsLinkComponent
Bulk GC supplies
Bulk GC supplies
Catalog # HEWL5190-3158
Supplier:  Agilent
Bulk GC supplies
Catalog # HEWL5190-3158
Supplier:  Agilent
Specifications
  • Description:
    Non-stick advanced green septa, 11 mm
  • Pk:
    400
Specifications
About this item
Agilent bulk chromatography supplies provide small, easily handled quantities in economical bulk packaging.
  • Economical and convenient packaging for bulk offerings
Ideal for high usage laboratories, Agilent bulk supplies provide the quality and consistency of Agilent chromatography supplies offered in convenient and economical packaging. Products available in bulk packaging include: inlet liners, gold inlet seals, liner o-rings, autosampler syringes, inlet septa, manual syringes, and autosampler vials.