Seals for GL screw caps, DURAN®
Silicone with PTFE coating.
- Thermal resistance: 130 °C (steam) and 200 °C (heat)
- Good chemical resistance thanks to the PTFE coating
Suitable for GL screw caps, premium and tamper-evident caps.
Frequently Bought Together