J&W DB-ALC1 and DB-ALC2 GC columns
: Agilent
DB-ALC1 and DB-ALC2 results are complete in just two minutes, reducing run time by one-third compared to other blood alcohol analysis columns.
- Reliable blood alcohol analysis
- Optimised primary and confirmation column pair for U.S. blood alcohol analysis
- Faster GC run times
- Available in 0,32 and 0,53 mm Int.Ø
Today’s automated static headspace systems use multi-sample equilibration ovens. Its unique polarity, DB-ALC2 provides better resolution of the key ethanol/acetone peaks than other available blood alcohol confirmation capillary columns. These columns are suitable for both headspace analysis and direct injections.
Frequently Bought Together