J&W DB-608 GC columns
: Agilent
DB-608 is specifically designed for the analysis of chlorinated pesticides and PCBs and is suitable for use in EPA methods 608, 508, and 8080.
- Specifically designed for the analysis of chlorinated pesticides and PCBs
- U.S. EPA Methods: 608, 508, 8080
- Excellent inertness and recoveries without pesticide breakdown
- Bonded and cross-linked
- Solvent rinsable
J&W DB-608 columns are precision-engineered to deliver industry-leading performance in the effective analysis of chlorinated pesticides and PCBs. DB-608 widely used for U.S. EPA methods 608, 508, and 8080, and deliver excellent inertness and recoveries, without resulting in pesticide breakdown. The columns serve as exact replacements for HP-608 and are bonded, cross-linked, and solvent rinsable. DB-608 also feature operating temperature limits of 40 to 260/300 °C and are tested to some of the strictest industry QC standards to ensure the highest levels of quality every time.
Frequently Bought Together