Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-(+)-Camptothecin
(S)-(+)-Camptothecin 2 mM
(S)-(+)-Camptothecin 2 mM
(S)-(+)-Camptothecin 2 mM
Catalog # API-02
Supplier:
G-Biosciences
CAS Number:
Expand image
(S)-(+)-Camptothecin 2 mM
Catalog # API-02
Supplier:
G-Biosciences
CAS Number:
Specifications
Pk:
1 mL
Seq:
2070
Specifications
About this item
Product Details
Documentation
:
:
348.36 g/mol
CAS Number:
7689-03-4
Formula:
C₂₀H₁₆N₂O₄
MDL Number:
MFCD00081076
Product Details
Documentation