Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Doxorubicin
Doxorubicin 2 mM
Doxorubicin 2 mM
Catalog # API-05
Supplier:
G-Biosciences
CAS Number:
Expand image
Doxorubicin 2 mM
Catalog # API-05
Supplier:
G-Biosciences
CAS Number:
Specifications
Pk:
50 µl
Seq:
1202
Specifications
About this item
Product Details
Documentation
:
:
543.52 g/mol
CAS Number:
23214-92-8
Formula:
C₂₇H₂₉NO₁₁
Product Details
Documentation