Murashige shoot tip rooting medium
Murashige shoot tip rooting medium 10×1 L, 10
Catalog # APOSPMM527-10X1L
Supplier: Apollo Scientific
Murashige shoot tip rooting medium
Catalog # APOSPMM527-10X1L
Supplier: Apollo Scientific
CAS Number:
Jump to:
Product Details & Documents
Contains macro/micronutrients, sucrose, agar andorganics as described by Malmgren, 1996 Plant tissue culture testedFull specification given in appendix.
- MFCD00243110
Specifications
Specifications
- Pk:10
- Description:Murashige shoot tip rooting medium 10×1 L