HRP - conjugate stabiliser, SeramunStab® STM
Catalog # APOSBIL3114-100ML
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:100 mL
- Description:SeramunStab® STM HRP - Conjugate stabiliser
- Seq:01
Specifications
About this item
Liquid product orange to red odourless pH 6,4 to 6,6. Stabiliser for conjugates with monomeric HRP. Preservation of very low conjugate concentrations.