Orders LinkContactUsLinkComponent
Capillary and fitting kits
Supplier:   Agilent
Capillary and fitting kits
Supplier:  Agilent
Certificates
Capillary and fitting kits
2 Options Available Below
Product Details & Documents
These starter kits contain the most widely used capillary tubing, Swagelok connectors, and fittings in a variety of sizes.
  • Designed-in for highest chromatographic performance
  • High quality products for lowest instrument downtime and best total cost of ownership
  • OEM products fit tightest Agilent specifications for validated environments
We’ve also included our flexible stainless steel capillaries to help you make the best LC connections, regardless of equipment brand. Plus, as a special bonus, all multi-use kits (HEWL5065-9938, HEWL5065-9937, HEWL5065-9939) feature a FREE cybertool that puts more than 30 lab essentials at fingertips. For high-efficiency columns, it’s best to use narrow-diameter red tubing (Int.Ø 0,12 mm), instead of conventional green (Int.Ø 0,17 mm) tubing.
Product Family Options
Product Information
  • Availability
    Price
  • HEWLG1316-68716
    1200 capillary kit for Int.Ø 0,12 mm
    1200 capillary kit for Int.Ø 0,12 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 410,00 €
    1 410,00 €
    1 * 1 ST
       1 410,00 €
  • HEWL5065-9947
    Rapid resolution HT capillary kit
    Rapid resolution HT capillary kit
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 766,00 €
    766,00 €
    1 * 1 ST
       766,00 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.