Graphite tubes and platforms
About this item
Graphite tubes are quality tested to ensure each batch passes our demanding performance specifications for contamination, sensitivity, precision, electrical resistance and lifetime.
- High purity graphite reduces spurious absorption from trace contaminants and improves signal-to-noise
- Every tube is hand-checked for reproducible and reliable results
- Perform fewer repeats and increase productivity
Use partitioned tubes for general purpose work and when you need the best detection limits with samples that have low background. These are also ideal when measuring organic solutions, or samples with low viscosity that may spread throughout the tube, potentially degrading precision.
Omega platform tubes combine ease-of-use with perfect atomisation: the integrated platform ensures the perfect thermal profile to produce dense atom clouds for good signal-to-noise ratios and the best detection limits. The combination of the solid pyrolytic platforms and plateau tubes provide continuity with older methods, and gives the best platform effect. This ensures best performance with samples that have high background or interferences.
3 Options Available Below
Product Details & Documents
Graphite tubes are quality tested to ensure each batch passes our demanding performance specifications for contamination, sensitivity, precision, electrical resistance and lifetime.
- High purity graphite reduces spurious absorption from trace contaminants and improves signal-to-noise
- Every tube is hand-checked for reproducible and reliable results
- Perform fewer repeats and increase productivity
Use partitioned tubes for general purpose work and when you need the best detection limits with samples that have low background. These are also ideal when measuring organic solutions, or samples with low viscosity that may spread throughout the tube, potentially degrading precision.
Omega platform tubes combine ease-of-use with perfect atomisation: the integrated platform ensures the perfect thermal profile to produce dense atom clouds for good signal-to-noise ratios and the best detection limits. The combination of the solid pyrolytic platforms and plateau tubes provide continuity with older methods, and gives the best platform effect. This ensures best performance with samples that have high background or interferences.
Product Family Options
Product Information
- AvailabilityPrice
- VARI6310001100Plateau tube, pyrolytically coated, for pyrolytic platforms onlyPlateau tube, pyrolytically coated, for pyrolytic platforms onlyLog in for availability1 * 1 ST786,69 €
- 636-2501Partitioned tubes, pyrolytically coated
- VARI6310001300Solid pyrolytic graphite platforms, only for plateau tubes