ContactUsLinkComponent
Distilling flask with side tube, Assistent
Distilling flask with side tube, Assistent
Artikelnr. HECH41037016
undefined
Distilling flask with side tube, Assistent
Artikelnr. HECH41037016
Spezifikationen
  • Capacity:
    250 ml
  • Pk:
    1
Spezifikationen
Über diesen Artikel
This distilling flask is made of borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
The length of the side-tube is approximately 200 mm.