Order Entry
Austria
Orders LinkContactUsLinkComponent
miRNA reference kit

miRNA reference kit

Universal miRNA reference kit, 10 ug, 1 KIT
Catalog # AGLS750700
Supplier:   AGILENT
miRNA reference kit
Catalog # AGLS750700
Supplier:  AGILENT
CAS Number:  
miRNA reference kit

Universal miRNA reference kit, 10 ug, 1 KIT

CAS Number:  
Price
684,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

The Universal Human miRNA Reference RNA is an ideal reference control for miRNA microarray or miRNA-targeted QRTPCR experiments.

  • Contains extensive range of miRNAs for genome-wide detection
  • Includes mRNA, large intergenic non-coding RNA, and piwi-interacting RNA in addition to miRNA
  • Non-synthetic, not limited to previously characterized miRNA species
  • Carefully screened for contaminating genomic DNA
  • Created in extremely large lots with stringent QC procedures, ensuring long-term availability of consistent reference RNA material

The Universal Human miRNA Reference RNA may also be used as an optimization or standardization reagent for these or other applications aimed at human miRNA analysis.

It is composed of total RNA from nine human tissues or cell lines specifically selected and formulated for optimal miRNA representation.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Universal miRNA reference kit, 10 ug

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.