ISIS supplies for 7500
Supplier: Agilent
Certificates
About this item
High quality supplies and components for the Integrated Sample Introduction System (ISIS) for use with the 7500 ICP-MS range.
- ETFE sample tubing with a 0,8 mm Int.Ø and 1,6 mm Ext.Ø is also available along with PTFE sample tubing with a 2 mm Int.Ø and 3 mm Int.Ø
- Alternatively, PFA sample tubing with 0,3 mm Int.Ø and 1,6 mm Ext.Ø can be provided
- A variety of joints are available in tee, cross or union form
- PTFE nuts and front and back ferrules for either 3 mm or 1,6 mm Ext.Ø can be provided
- Spiral tubing, peristaltic pump tubing adapters and tubing clamps, in small or large models, may be purchased
- Wrenches for valve maintenance and tags for tubing identification may also be provided
- A range of standard accessory parts for high sample throughput, auto dilution or discrete sampling are provided
Agilent's unique ISIS enhances the functionality and flexibility of the Agilent 7500 Series ICP-MS. ISIS features high precision peristaltic pumps and an optional switching valve to allow complete flexibility of configuration. The ISIS hardware is built into the 7500 chassis and close-coupled to increase productivity through high-speed sample uptake and rapid washout. To enhance the capabilities that the system offers and maintain performance even after heavy use, Agilent provides a range of replacement components and other supplies.
Frequently Bought Together