Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Chee and pool vitis Cd2 medium

Chee and pool vitis Cd2 medium 10×1 L, 10
Catalog # APOSPMC287-10X1L
Supplier:   Apollo Scientific
Chee and pool vitis Cd2 medium
Catalog # APOSPMC287-10X1L
Supplier:  Apollo Scientific
CAS Number:  
Chee and pool vitis Cd2 medium

Chee and pool vitis Cd2 medium 10×1 L, 10

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 10 ST
Product Details & Documents

Contains macro/micronutrients as describedby Chee and PoolPlant tissue culture testedFull specification given in appendix.

  • MFCD00282869
Specifications

Specifications

  • Pk:
    10
  • Description:
    Chee and pool vitis Cd2 medium 10×1 L

Specifications