ContactUsLinkComponent
HRP - conjugate stabiliser, SeramunStab® STC
HRP - conjugate stabiliser, SeramunStab® STC
Catalog # APOSBIL3115-100ML
Supplier:  Apollo Scientific
CAS Number:  
HRP - conjugate stabiliser, SeramunStab® STC
Catalog # APOSBIL3115-100ML
Supplier:  Apollo Scientific
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    SeramunStab® STC HRP - conjugate stabiliser
  • Seq:
    01
Specifications
About this item
Liquid product red odourless pH-value 7,2 to 7,4 boiling point 103 °C. Optimised for background reduction.