ContactUsLinkComponent
HRP-conjugate stabiliser, SeramunStab® STNP
HRP-conjugate stabiliser, SeramunStab® STNP
Catalog # APOSBIL3113-100ML
Supplier:  Apollo Scientific
CAS Number:  
HRP-conjugate stabiliser, SeramunStab® STNP
Catalog # APOSBIL3113-100ML
Supplier:  Apollo Scientific
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    SeramunStab® STNP HRP-conjugate stabiliser
  • Seq:
    01
Specifications
About this item
Liquid product red odourless pH 6,7 to 6,9. Protein free Stabiliser. Enzyme activity preservation by special chemistry.