ContactUsLinkComponent
Connecting capillary, VWD
Connecting capillary, VWD
Catalog # HEWLG1314-87302
Supplier:  Agilent
undefined
Connecting capillary, VWD
Catalog # HEWLG1314-87302
Supplier:  Agilent
Specifications
  • Ø int.:
    0,17 mm
  • Description:
    Capillary, S/E fittings, Stainless steel
  • Length:
    0,12 m
  • Pk:
    1
Specifications
About this item
A comprehensive range of capillaries and fittings are available to protect the integrity of the chromatographic results.
  • Best fit to detector flow cell for optimum chromatographic performance
  • Increased usability without scarifying durability
  • Tight and leak free connection