Glycan mapping HPLC columns, AdvanceBio
Supplier: Agilent
Certificates
About this item
AdvanceBio Glycan Mapping Columns for fast, reproducible, high resolution glycan characterisation.
- Speed: Obtain glycan maps in less than 10 minutes, superior results – in 40% less time than the competition
- High-resolution: Characterise both labeled and unlabeled glycans, and identify any glycosylation changes that occur due to process variables
- Flexibility: 2,7 µm superficially porous particles for high resolution with less backpressure. For UHPLC analysis 1,8 µm particles are an ideal choice
- Reproducibility: QA tested with a 2-AB labeled IgG N-linked glycan sample to ensure consistent quality and performance
AdvanceBio Glycan Mapping columns leverage technology that optimises results for MS and fluorescence detection. Choose from two UHPLC configurations: 2,7 µm superficially porous, for high resolution and lower backpressure, or 1,8 µm for highest resolution.
Order AdvanceBio Glycan Mapping columns with a selection of standards for performance testing and retention mapping of labeled and unlabeled glycans. Our AdvanceBio N-glycan sample preparation kits provide all the components needed for the sample prep workflow with a comprehensive procedure to simplify the process.
Frequently Bought Together