Orders LinkContactUsLinkComponent
Universal reference RNAs
Universal reference RNAs
Supplier:   AGILENT
Universal reference RNAs
Supplier:  AGILENT
Certificates
Universal reference RNAs
3 Options Available Below
Product Details & Documents
High-quality total RNA for human, mouse, and rat microarray gene-expression profiling that acts as a consistent control for standard data set comparisons.
  • Comprised of 10 cell lines for human, 11 for mouse and 14 for rat giving broad gene coverage
  • UHRR selected as one of the two standards in the FDA led Microarray Quality Control (MAQC) project
The reference RNA is designed to be used as a reference for uarray gene-profiling experiments.
Delivery information: Includes RNase-free water, 1,5 ml.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • AGLS740000
    Universal human reference RNA, 2×200 µg
    Universal human reference RNA, 2×200 µg
    2
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 1 033,00 €
    1 033,00 €
    1 * 2 ST
       1 033,00 €
  • AGLS740100
    Universal mouse reference RNA, 2×200 µg
    Universal mouse reference RNA, 2×200 µg
    2
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 1 012,00 €
    1 012,00 €
    1 * 2 ST
       1 012,00 €
  • AGLS740200
    Universal rat reference RNA, 2×200 µg
    Universal rat reference RNA, 2×200 µg
    2
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 1 012,00 €
    1 012,00 €
    1 * 2 ST
       1 012,00 €