Orders LinkContactUsLinkComponent
J.T.Baker®, Plate 96
J.T.Baker®, Plate 96
   Sustainable Options Available
Supplier:   Avantor
J.T.Baker®, Plate 96
Supplier:  Avantor
   Sustainable Options Available
Certificates
J.T.Baker®, Plate 96
4 Options Available Below
Product Details & Documents
J.T.Baker Plate 96 round well plates made from PS are ideal for ELISA applications.
  • Alphanumeric coding assists with identification of samples
  • Plate shape significantly reduces sample carry-over
  • Standardised dimensions (128×85 mm) ensure compatibility with most common laboratory machines
Specifications for Product Family
Specifications
  • Catalog No:
  • 43001-0117
  • 43001-0118
  • 43001-0119
  • 43001-0600
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
Specifications
Product Family Options
Product Information
  • Colour
    Sterile
    Well volume
    Availability
    Price
  • 43001-0117
    PS plates, U-bottom
    PS plates, U-bottom
    Clear
    -
    0,3 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 106,08 €
    106,08 €
    1 * 100 ST
       106,08 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0117
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 43001-0118
    PS plates, V-bottom
    PS plates, V-bottom
    Clear
    -
    0,24 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 106,09 €
    106,09 €
    1 * 100 ST
       106,09 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0118
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 43001-0119
    PS plates, F-bottom
    PS plates, F-bottom
    Clear
    -
    0,35 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 106,08 €
    106,08 €
    1 * 100 ST
       106,08 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0119
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 43001-0600
    PP plates, U-bottom
    PP plates, U-bottom
    Transparent
    -
    0,3 ml
    Log in for availability
    Log in for availability
    1 * 64 ST
    1 * 64 ST - 68,09 €
    68,09 €
    1 * 64 ST
       68,09 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0600
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
      Low Manufacturing Impact