ContactUsLinkComponent
J.T.Baker®, Robotic Tips Type QG
J.T.Baker®, Robotic Tips Type QG
  49018-0106
 :  Avantor
   Low Manufacturing Impact
undefined
J.T.Baker®, Robotic Tips Type QG
  49018-0106
 :  Avantor
   Low Manufacturing Impact
  • Packed:
    12× 2 racks, 96 each, blistered
  • Sterile:
    -
  • Volume:
    50 µl
  • Seq:
    1
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • Tip colour:
    Black
  • Filter:
    +
  • Pk:
    2.304
Suitable for use on third party robotic instruments QIAgility. Available in conductive version, with filter, non-sterile, and in CE-IVD marked version for clinical diagnostic applications.
  • Manufactured under cleanroom conditions (cleanroom class 8)
  • State of the art quality control
  • Free of DNA, DNase, RNase and PCR inhibitors
  • Ideal for high-throughput analysis and DNA purification
  • Packed in blister
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Tips also suitable for several Tecan instruments