Order Entry
Austria
ContactUsLinkComponent
Total RNA isolation mini kit
Total RNA isolation mini kit
Catalog # AGLS5185-6000
Supplier:  AGILENT
CAS Number:  
Total RNA isolation mini kit
Catalog # AGLS5185-6000
Supplier:  AGILENT
CAS Number:  

Specifications

  • Pk:
    1 kt
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    Total RNA isolation mini kit
  • Description:
    Total RNA isolation mini kit

Specifications

About this item

RNA isolation kits provide a straightforward, spin-column method to deliver RNA for use in gene expression and other downstream analysis.