Order Entry
Austria
Orders LinkContactUsLinkComponent
 

HEPES Buffer Solution 1 M, HyClone™

HyClone HEPES 1M Solution, Glass bottle, 100 ml
Catalog # HYCLSH30237.01
HEPES Buffer Solution 1 M, HyClone™
Catalog # HYCLSH30237.01
CAS Number:  
HEPES Buffer Solution 1 M, HyClone™
HEPES Buffer Solution 1 M, HyClone™

HyClone HEPES 1M Solution, Glass bottle, 100 ml

CAS Number:  
Price
23,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Maintain physiological pH in cell culture, despite changes in carbon dioxide concentration (produced by cellular respiration) with Cytiva HyClone HEPES solution.

  • 0,1 µm sterile filtered
  • pH: 7,2 - 7,5
  • Storage: 2 to 8° C

 : Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities

 

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Description:
    HyClone HEPES 1M Solution

Specifications