Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(-)-Luciferin sodium salt
D(-)-Luciferin sodium salt ≥99.5% endotoxin-free
D(-)-Luciferin sodium salt ≥99.5% endotoxin-free
OZBILN10000
:
OZ BIOSCIENCES
:
D(-)-Luciferin sodium salt ≥99.5% endotoxin-free
OZBILN10000
:
OZ BIOSCIENCES
:
Pk:
1 g
Seq:
3047
:
:
302.30 g/mol
:
103404-75-7
:
C₁₁H₇N₂NaO₃S₂
:
MFCD00044938