Order Entry
Austria
ContactUsLinkComponent
 
J.T.Baker® BAKER INSTRA-ANALYZED®, Platinum Plasma Standards
  5740.0100
 :  Avantor
 :  
J.T.Baker® BAKER INSTRA-ANALYZED®, Platinum Plasma Standards
  5740.0100
 :  Avantor
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    10% HCl
  • Pk:
    100 mL
  • Seq:
    2
  • Element:
    Platinum
  • Symbol:
    Pt