Order Entry
Austria
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Manganese Plasma Standard
J.T.Baker® BAKER INSTRA-ANALYZED®, Manganese Plasma Standard
Catalog # 5735.0100
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKER INSTRA-ANALYZED®, Manganese Plasma Standard
Catalog # 5735.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Element:
    Manganese
  • Symbol:
    Mn

Specifications

About this item