Order Entry
Austria
Orders LinkContactUsLinkComponent
 

J.T.Baker® DILUT-IT™, Buffer Solutions, Concentrate

Supplier:   Avantor
J.T.Baker® DILUT-IT™, Buffer Solutions, Concentrate
Supplier:  Avantor
Certificates
J.T.Baker® DILUT-IT™, Buffer Solutions, Concentrate

2 Options Available Below

Product Details & Documents

Volumetric concentrate, for dilution to 500 ml.

  • Traceable to NIST
Product Family Options

Product Information

  • pH value
    pH accuracy
    pH buffer type
    Reference temperature
    Pk
    Availability
    Price
  • 4795.
    DILUT-IT™ concentrate
    DILUT-IT™ concentrate
    4,00
    ±0,01
    Phthalate
    25 °C
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,73 €
    37,73 €
    1 * 1 ST
       37,73 €

  • 4796.
    DILUT-IT™ concentrate
    DILUT-IT™ concentrate
    7,00
    ±0,01
    Phosphate
    25 °C
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,47 €
    38,47 €
    1 * 1 ST
       38,47 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.