Order Entry
Austria
Orders LinkContactUsLinkComponent
J.T.Baker® DILUT-IT™, Buffer Solutions, Colour Coded, Concentrate

J.T.Baker® DILUT-IT™, Buffer Solutions, Colour Coded, Concentrate

Supplier:   Avantor
J.T.Baker® DILUT-IT™, Buffer Solutions, Colour Coded, Concentrate
Supplier:  Avantor
Certificates
J.T.Baker® DILUT-IT™, Buffer Solutions, Colour Coded, Concentrate

2 Options Available Below

Product Details & Documents

Volumetric concentrate, for dilution to 500 ml.

  • Traceable to NIST

Documents

Certificates
Search by Lot Number
Product Family Options

Product Information

  • pH value
    pH accuracy
    pH buffer type
    Colour
    Pk
    Availability
    Price
  • 4861.
    DILUT-IT™ concentrate
    DILUT-IT™ concentrate
    7,00
    ±0,02
    Phosphate
    Green
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,17 €
    49,17 €
    1 * 1 ST
       49,17 €

  • 4862.
    DILUT-IT™ concentrate
    DILUT-IT™ concentrate
    9,00
    ±0,02
    Borate
    Blue
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST