Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Human recombinant Set7/9 Histone Methyltransferase
Human recombinant Set7/9 Histone Methyltransferase
APOSBITP1326-1MG
:
Apollo Scientific
:
Human recombinant Set7/9 Histone Methyltransferase
APOSBITP1326-1MG
:
Apollo Scientific
:
Pk:
1 mg
Enzyme type:
Recombinant
Species:
Human
Storage conditions:
Ambient
Enzyme name:
Set7/9 Histone Methyltransferase
Enzyme synonyms:
HMTase