Macerozyme R-10
A mixture of pectinase, cellulase and hemicellulase
- Storage conditions:Freezer
- MDL No.:MFCD00131809
- Enzyme name:Macerozyme R-10
- Enzyme synonyms:Pectinase
Frequently Bought Together
A mixture of pectinase, cellulase and hemicellulase