Order Entry
Austria
ContactUsLinkComponent
Anti-IL-4 Mouse Antibody
Anti-IL-4 Mouse Antibody
Catalog # APOSBITP1979-1MG
Supplier:  Apollo Scientific
Anti-IL-4 Mouse Antibody
Catalog # APOSBITP1979-1MG
Supplier:  Apollo Scientific

Specifications

  • Antigen name:
    Interleukin-4
  • Antigen symbol:
    IL-4
  • Host:
    Mouse
  • Reactivity:
    Human
  • Pk:
    1 mg

Specifications

About this item