Order Entry
Austria
ContactUsLinkComponent
 
Human recombinant FGF-2 (147)
Catalog # APOSBITP2078-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant FGF-2 (147)
Catalog # APOSBITP2078-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    23
  • Protein synonyms:
    Fibroblast Growth Factor-basic (147 a.a)
  • Protein/peptide name:
    FGF-2 (147)

Specifications

About this item