Order Entry
Austria
ContactUsLinkComponent
Recombinant Japanese Encephalitis Virus
Recombinant Japanese Encephalitis Virus
Catalog # APOSBITP1925-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Japanese Encephalitis Virus
Catalog # APOSBITP1925-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    Encephalitis Japanese
  • Protein/peptide name:
    Japanese Encephalitis Virus

Specifications

About this item