Order Entry
Austria
ContactUsLinkComponent
 
Recombinant HTLV-1 gp46
Catalog # APOSBITP1911-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant HTLV-1 gp46
Catalog # APOSBITP1911-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    HTLV-1 gp46

Specifications

About this item