Order Entry
Austria
ContactUsLinkComponent
Recombinant HIV-1 gag p17, p24, gp120
Recombinant HIV-1 gag p17, p24, gp120
Catalog # APOSBITP1889-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant HIV-1 gag p17, p24, gp120
Catalog # APOSBITP1889-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HIV-1 p17/24/120
  • Protein/peptide name:
    HIV-1 gag p17, p24, gp120

Specifications

About this item