Order Entry
Austria
ContactUsLinkComponent
Recombinant HIV-1 gp41, Biotin
Recombinant HIV-1 gp41, Biotin
Catalog # APOSBITP1875-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant HIV-1 gp41, Biotin
Catalog # APOSBITP1875-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    HIV-1 gp41

Specifications

About this item