Recombinant HIV-1 p24, HRP (Horseradish Peroxidase)
Supplier: Apollo Scientific
Certificates
About this item
Specifications
- Conjugation:HRP (Horseradish Peroxidase)
- Protein/peptide type:Recombinant
- Protein synonyms:HIV-1 24
- Protein/peptide name:HIV-1 p24
Specifications
Frequently Bought Together