Order Entry
Austria
ContactUsLinkComponent
Recombinant HIV-1 p24, Biotin
Recombinant HIV-1 p24, Biotin
Catalog # APOSBITP1870-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant HIV-1 p24, Biotin
Catalog # APOSBITP1870-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    rHIV-1 p24
  • Protein/peptide name:
    HIV-1 p24

Specifications

About this item