Order Entry
Austria
ContactUsLinkComponent
Recombinant Cytomegalo Virus gB
Recombinant Cytomegalo Virus gB
Catalog # APOSBITP1839-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Cytomegalo Virus gB
Catalog # APOSBITP1839-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    CMV gB
  • Protein/peptide name:
    Cytomegalo Virus gB

Specifications

About this item