0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Proteins and Peptides
Recombinant Hepatitis E Virus ORF3
Recombinant Hepatitis E Virus ORF3
APOSBITP1831-1MG
:
Apollo Scientific
:
Recombinant Hepatitis E Virus ORF3
APOSBITP1831-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
HEV ORF3
Protein/peptide name:
Hepatitis E Virus ORF3