Order Entry
Austria
ContactUsLinkComponent
Recombinant Hepatitis B Virus HBe
Recombinant Hepatitis B Virus HBe
Catalog # APOSBITP1824-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis B Virus HBe
Catalog # APOSBITP1824-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HBV HBe
  • Protein/peptide name:
    Hepatitis B Virus HBe

Specifications

About this item